Learn More
TARGETMOL CHEMICALS INC LY 456236 HYDROCHLORIDE 5MG
SDP
Catalog No. 502580908
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LY 456236 hydrochloride (MPMQ hydrochloride) is an mGlu1 receptor antagonist with an ic50 value of 143 nM. LY456236 has selective non-competitive and oral activity and can inhibit the hydrolysis of inositol phosphate with IC50 of 0.145 uM. LY 456236 hydrochloride inhibited EGFR with IC50 of 0.91 uM. purity: 99%
Catalog No. 50-258-0908 Supplier TARGETMOL CHEMICALS INC Supplier No. T229545MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LY 456236 hydrochloride (MPMQ hydrochloride) is an mGlu1 receptor antagonist with an ic50 value of 143 nM. LY456236 has selective non-competitive and oral activity and can inhibit the hydrolysis of inositol phosphate with IC50 of 0.145 uM. LY 456236 hydrochloride inhibited EGFR with IC50 of 0.91 uM. purity: 99%