Learn More
STEMCELL Technologies Anti-Mouse Ly-6G Antibody, Clone 1A8, Pacific Blue, Size: 25 ug
SDP
Supplier:  STEMCELL Technologies 60031PB.1
Encompass
Clone 1A8 is a rat monoclonal IgG2a antibody that reacts with mouse Ly-6G a 21 - 25 kDa GPI-anchored protein which together with the structurally related Ly-6C protein comprises the granulocyte receptor-1 antigen. Verified for use with flow cytometry assays use this antibody in applications requiring the detection of Ly-6G. The 1A8 antibody binds specifically to Ly-6G whereas another commonly used antibody clone RB6-8C5 binds to both Ly-6G and Ly-6C. It has been reported that the 1A8 antibody detects Ly-6G-expressing granulocytes in peripheral blood whereas the RB6-8C5 antibody also binds to Ly-6C-expressing lymphocytes monocytes and dendritic cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep kits including EasySep Mouse Neutrophil Enrichment Kit (Catalog 19762). Visit STEMCELL.com for more information and related products.
Catalog No. 50-239-0379