Learn More
Enzo Life Sciences LY-83,583 (5mg). CAS: 91300-60-6
SDP
Supplier:  Enzo Life Sciences ALX550002M005
Encompass_Preferred
Competitive inhibitor of soluble guanylyl cyclase (sGC) (IC50=2µM) and of cGMP production. Neurotoxin. Blocks antigen-induced leukotriene release and reduces platelet-endothelial cell adhesion. Blocks acethylcholine-induced vasorelaxation. Alternative name: 6-(Phenylamino)-5,8-quinolinedione, 6-Anilino-5,8-quinolinedione. Purity: ≥95% (TLC). Appearance: Violet to dark red powder. Solubility: Soluble in methanol, methylene chloride or diluted aqueous acid; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9455