Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LY117018 TFA 25mg | 2390041-98-0 | 25 MG
SDP

Catalog No. 5000228232
Encompass_Preferred

LY117018 TFA is a raloxifene analog and a selective estrogen receptor modulator (SERM) reported to exert antiproliferative effects on breast cancer cell lines. Supplied as a trifluoroacetate salt, the compound is provided as a small-scale research reagent for in vitro studies.

  • Selective estrogen receptor modulator activity.
  • Reported antiproliferative effects on breast cancer cell lines.
  • Supplied as a trifluoroacetate salt for improved handling.
  • Available in milligram-scale quantities for research use.
  • Chemical formula C29H26F3NO6S; molecular weight 573.58 g/mol.

Catalog No. 50-002-28232 Supplier Medchemexpress LLC Supplier No. HY116896A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY117018 TFA is a raloxifene analog and a selective estrogen receptor modulator (SERM) reported to exert antiproliferative effects on breast cancer cell lines. Supplied as a trifluoroacetate salt, the compound is provided as a small-scale research reagent for in vitro studies.

  • Selective estrogen receptor modulator activity.
  • Reported antiproliferative effects on breast cancer cell lines.
  • Supplied as a trifluoroacetate salt for improved handling.
  • Available in milligram-scale quantities for research use.
  • Chemical formula C29H26F3NO6S; molecular weight 573.58 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.