Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LY117018 TFA 5mg | 2390041-98-0 | 573.58 | C29H26F3NO6S | 5 MG
SDP

Catalog No. 5000226216
Encompass_Preferred

LY117018 TFA is the trifluoroacetic acid salt of LY117018, a raloxifene analog and selective estrogen receptor modulator reported to exert antiproliferative effects on breast cancer cell lines. It is provided as a research reagent for in vitro and in vivo studies of estrogen receptor signaling and related pathways.

  • Selective estrogen receptor modulator with reported antiproliferative activity.
  • Soluble in DMSO up to 100 mg/mL; sonication may be required.
  • In vivo formulations reported with solubility ≈5 mg/mL using common co-solvent systems.
  • Recommended storage: solid sealed and kept away from moisture; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Molecular formula C29H26F3NO6S; molecular weight 573.58.

Catalog No. 50-002-26216 Supplier Medchemexpress LLC Supplier No. HY116896A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY117018 TFA is the trifluoroacetic acid salt of LY117018, a raloxifene analog and selective estrogen receptor modulator reported to exert antiproliferative effects on breast cancer cell lines. It is provided as a research reagent for in vitro and in vivo studies of estrogen receptor signaling and related pathways.

  • Selective estrogen receptor modulator with reported antiproliferative activity.
  • Soluble in DMSO up to 100 mg/mL; sonication may be required.
  • In vivo formulations reported with solubility ≈5 mg/mL using common co-solvent systems.
  • Recommended storage: solid sealed and kept away from moisture; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Molecular formula C29H26F3NO6S; molecular weight 573.58.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.