Learn More
Selleck Chemical LLC LY2109761 S2704-200mg
SDP
Catalog No. 503110346
Encompass_Preferred
LY2109761 is a novel selective TGF- receptor type I/II (T RI/II) dual inhibitor with Ki of 38 nM and 300 nM in a cell-free assay respectively shown to negatively affect the phosphorylation of Smad2 LY2109761 blocks autophagy and induces apoptosis
Catalog No. 50-311-0346 Supplier Selleck Chemical LLC Supplier No. S2704200MG
LY2109761 is a novel selective TGF- receptor type I/II (T RI/II) dual inhibitor with Ki of 38 nM and 300 nM in a cell-free assay respectively shown to negatively affect the phosphorylation of Smad2 LY2109761 blocks autophagy and induces apoptosis