Learn More
Selleck Chemical LLC LY2119620 5mg 886047-22-9
SDP
Supplier:  Selleck Chemical LLC S73665MG
Encompass_Preferred
LY2119620 is a specific, and allosteric agonist of human M2 and M4 muscarinic acetylcholine receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8191