Learn More
Apexbio Technology LLC LY2157299(Synonyms: Galunisertib, LY-2157299, LY2157299 monohydrate, LY2157299 hydrate, LY2157299 free base), 10mM (in 1mL DMSO), CAS: 700874-72-2.
SDP
Catalog No. 5000060937
Encompass_Preferred
LY2157299 (CAS 700874-72-2) is a small-molecule antagonist that selectively targets the transforming growth factor-beta type I receptor (T RI) By blocking signaling through T RI it prevents phosphorylation of downstream effectors Smad2 and Smad3 thereby interfering with the TGF- signaling pathway Preclinical studies indicate LY2157299 inhibits 1-integrin activation reduces tumor cell invasion lowers connective tissue growth factor (CTGF) production and suppresses angiogenesis Currently in phase II clinical trials LY2157299 is being investigated primarily for therapeutic potential against hepatocellular carcinoma and glioblastoma
Catalog No. 50-000-60937 Supplier Apexbio Technology LLC Supplier No. A83485.1
LY2157299 (CAS 700874-72-2) is a small-molecule antagonist that selectively targets the transforming growth factor-beta type I receptor (T RI) By blocking signaling through T RI it prevents phosphorylation of downstream effectors Smad2 and Smad3 thereby interfering with the TGF- signaling pathway Preclinical studies indicate LY2157299 inhibits 1-integrin activation reduces tumor cell invasion lowers connective tissue growth factor (CTGF) production and suppresses angiogenesis Currently in phase II clinical trials LY2157299 is being investigated primarily for therapeutic potential against hepatocellular carcinoma and glioblastoma