Learn More
Selleck Chemical LLC Galunisertib (LY2157299) 5mg 700874-72-2
SDP
Supplier:  Selleck Chemical LLC S22305MG
Encompass_Preferred
Galunisertib (LY2157299) is a potent TGF? receptor I (T?RI) inhibitor with IC50 of 56 nM in a cell-free assay. Phase 2/3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0253456