Learn More
Cayman Chemical LY2183240 2’-isomer, Purity: ≥98%, CAS: 1010096-65-7, 5 mg
SDP
Catalog No. 502968940
Encompass_Preferred
A less potent, 2,5-regioisomer of LY2183240 that inhibits anandamide hydrolysis and uptake with IC50 values of 33 and 998 nM, respectively
Catalog No. 50-296-8940 Supplier Cayman Chemical Supplier No. 145235 MG
A less potent, 2,5-regioisomer of LY2183240 that inhibits anandamide hydrolysis and uptake with IC50 values of 33 and 998 nM, respectively