Learn More
Apexbio Technology LLC LY2228820 862507-23-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061592
Encompass_Preferred
LY2228820 (CAS 862507-23-1) is a potent selective ATP-competitive small-molecule inhibitor targeting the and isoforms of p38 mitogen-activated protein kinase (MAPK) with reported IC50 values of 5 3 nM and 3 2 nM respectively By suppressing p38 MAPK signaling LY2228820 decreases phosphorylation of heat shock protein 27 (HSP27) induced by Bortezomib thereby elevating cytotoxic effects in multiple myeloma (MM) models Additionally it inhibits secretion of IL-6 by bone marrow mononuclear cells and stromal cells as well as secretion of MIP-1 by patient-derived MM cell subsets and osteoclast precursors In melanoma (B16-F10) and NSCLC (A549) xenograft murine models LY2228820 reduces phosphorylation of MK2 and slows tumor progression respectively highlighting its potential utility in oncology research
Catalog No. 50-000-61592 Supplier Apexbio Technology LLC Supplier No. A55665.1
LY2228820 (CAS 862507-23-1) is a potent selective ATP-competitive small-molecule inhibitor targeting the and isoforms of p38 mitogen-activated protein kinase (MAPK) with reported IC50 values of 5 3 nM and 3 2 nM respectively By suppressing p38 MAPK signaling LY2228820 decreases phosphorylation of heat shock protein 27 (HSP27) induced by Bortezomib thereby elevating cytotoxic effects in multiple myeloma (MM) models Additionally it inhibits secretion of IL-6 by bone marrow mononuclear cells and stromal cells as well as secretion of MIP-1 by patient-derived MM cell subsets and osteoclast precursors In melanoma (B16-F10) and NSCLC (A549) xenograft murine models LY2228820 reduces phosphorylation of MK2 and slows tumor progression respectively highlighting its potential utility in oncology research