Learn More
STEMCELL Technologies LY2228820 (Dimesylate), Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 74162
Encompass
LY2228820 is a trisubstituted imidazole derivative and a potent inhibitor of the α- and β-isoforms of p38 MAP kinase (MAPK) in vitro (IC₅₀ = 5.3 and 3.2 nM, respectively) with anti-inflammatory and anti-neoplastic activities (Campbell et al.). This product is supplied as the dimesylate salt of the molecule.MAINTENANCE AND SELF-RENEWAL· Combined with other small molecule inhibitors such as Rapamycin (Catalog #73362) and SR1 (Catalog #72342; Li et al.) or SB203580 (Catalog #72222), Vx702, and BIRB-796 (Catalog #72682), enhances the self-renewal of cord blood-derived hematopoietic stem cells (Baudet et al.).CANCER RESEARCH· By inhibiting p38 MAPK, which is highly expressed in human cancers, LY2228820 is potent and selective at inhibiting tumor growth in animal models of a variety of human cancers (Campbell et al.).
Catalog No. 50-197-6191