Learn More
Selleck Chemical LLC Ralimetinib dimesylate 50mg 862507-23-1 LY2228820 dimesylate
SDP
Catalog No. 508034
Encompass_Preferred
Ralimetinib (LY2228820) dimesylate is a novel and potent inhibitor of p38 MAPK with IC50 of 7 nM in a cell-free assay, does not alter p38 MAPK activation. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-803-4 Supplier Selleck Chemical LLC Supplier No. S149450MG
Ralimetinib (LY2228820) dimesylate is a novel and potent inhibitor of p38 MAPK with IC50 of 7 nM in a cell-free assay, does not alter p38 MAPK activation. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.