Learn More
Selleck Chemical LLC Ralimetinib dimesylate 5mg 862507-23-1 LY2228820 dimesylate
SDP
Catalog No. NC0432312
Encompass_Preferred
Ralimetinib (LY2228820) dimesylate is a novel and potent inhibitor of p38 MAPK with IC50 of 7 nM in a cell-free assay, does not alter p38 MAPK activation. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0432312 Supplier Selleck Chemical LLC Supplier No. S14945MG
Add to Cart
Add to Cart
Ralimetinib (LY2228820) dimesylate is a novel and potent inhibitor of p38 MAPK with IC50 of 7 nM in a cell-free assay, does not alter p38 MAPK activation. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.