Learn More
Cayman Chemical LY2228820 (mesylate), Purity: ≥98%, CAS: 862507-23-1, 10 mg
SDP
Catalog No. 502986856
Encompass_Preferred
A potent inhibitor of p38α MAP kinase (IC50 = 7 nM for human recombinant p38α); inhibits LPS-induced TNF-α production in murine peritoneal macrophages and reduces phosphorylation of MK2 in RAW 264.7 cells (IC50s = 5.2 and 34.3 nM, respectively); inhibits TNF-α production in LPS-treated mice and reduces paw swelling and cartilage destruction in a rat model of chronic inflammation (TMED50s = <1, 1.5, and 1.5 mg/kg, respectively)
Catalog No. 50-298-6856 Supplier Cayman Chemical Supplier No. 2325910 MG
A potent inhibitor of p38α MAP kinase (IC50 = 7 nM for human recombinant p38α); inhibits LPS-induced TNF-α production in murine peritoneal macrophages and reduces phosphorylation of MK2 in RAW 264.7 cells (IC50s = 5.2 and 34.3 nM, respectively); inhibits TNF-α production in LPS-treated mice and reduces paw swelling and cartilage destruction in a rat model of chronic inflammation (TMED50s = <1, 1.5, and 1.5 mg/kg, respectively)