Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ly2365109 hydrochloride | 1779796-27-8 | 98.7% | 421.91 | C22H28ClNO5 | 25 MG
SDP

Catalog No. 5000215056
Encompass_Preferred

LY2365109 hydrochloride is a potent, selective glycine transporter 1 (GlyT1) inhibitor that increases glycine levels in cerebrospinal fluid and raises seizure thresholds in preclinical models. It is provided as a solid research reagent with characterized purity and stability profiles.

  • Potent and selective GlyT1 inhibitor, IC50 15.8 nM for glycine uptake.
  • Elevates CSF glycine levels in preclinical studies.
  • Shown to increase seizure thresholds in animal models.
  • High purity suitable for research use (98.69%).
  • White to off-white solid, molecular weight 421.91, formula C22H28ClNO5.
  • Soluble in DMSO (≥31 mg/mL) and insoluble in water (<0.1 mg/mL).
  • Recommended storage sealed at 4°C; in solvent store at -80°C for long term.

Catalog No. 50-002-15056 Supplier Medchemexpress LLC Supplier No. HY100416A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY2365109 hydrochloride is a potent, selective glycine transporter 1 (GlyT1) inhibitor that increases glycine levels in cerebrospinal fluid and raises seizure thresholds in preclinical models. It is provided as a solid research reagent with characterized purity and stability profiles.

  • Potent and selective GlyT1 inhibitor, IC50 15.8 nM for glycine uptake.
  • Elevates CSF glycine levels in preclinical studies.
  • Shown to increase seizure thresholds in animal models.
  • High purity suitable for research use (98.69%).
  • White to off-white solid, molecular weight 421.91, formula C22H28ClNO5.
  • Soluble in DMSO (≥31 mg/mL) and insoluble in water (<0.1 mg/mL).
  • Recommended storage sealed at 4°C; in solvent store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.