Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ly2365109 hydrochloride | 1779796-27-8 | 98.7% | 421.91 | C22H28ClNO5 | 50 MG
SDP

Catalog No. 5000213908
Encompass_Preferred

LY2365109 hydrochloride is a potent, selective inhibitor of the glycine transporter 1 (GlyT1) used for laboratory research. It inhibits glycine uptake in hGlyT1a-expressing cells with an IC50 of 15.8 nM and is supplied as a white to off-white solid with high reported purity.

  • Potent, selective GlyT1 inhibition (IC50 15.8 nM).
  • High reported purity (98.7%).
  • White to off-white solid suitable for handling and formulation.
  • Provided with standard chemical identifiers (CAS 1779796-27-8; C22H28ClNO5; MW 421.91).
  • Available in multiple pack sizes, including 50 MG for laboratory use.

Catalog No. 50-002-13908 Supplier Medchemexpress LLC Supplier No. HY100416A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY2365109 hydrochloride is a potent, selective inhibitor of the glycine transporter 1 (GlyT1) used for laboratory research. It inhibits glycine uptake in hGlyT1a-expressing cells with an IC50 of 15.8 nM and is supplied as a white to off-white solid with high reported purity.

  • Potent, selective GlyT1 inhibition (IC50 15.8 nM).
  • High reported purity (98.7%).
  • White to off-white solid suitable for handling and formulation.
  • Provided with standard chemical identifiers (CAS 1779796-27-8; C22H28ClNO5; MW 421.91).
  • Available in multiple pack sizes, including 50 MG for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.