Learn More
Cayman Chemical LY2365109 (hydrochloride), Purity: ≥95%, CAS: 1779796-27-8, 50 mg
SDP
Catalog No. 502916164
Encompass_Preferred
An inhibitor of GlyT1a (IC50 = 15.8 nM in BE2-C cells expressing the human transporter); selective for GlyT1a over GlyT2 (IC50 = >30 µM in RD cells expressing the human transporter); induces locomotor hyperactivity, impaired gait, compulsive walking, and labored breathing, as well as impairs the ability to stand, in rats at 10 mg/kg; prevents maximal electroshock-induced acute seizures in mice at 30 mg/kg and decreases seizure number in an electroshock-induced rat model of chronic epilepsy at 10 mg/kg
Catalog No. 50-291-6164 Supplier Cayman Chemical Supplier No. 3742750 MG
An inhibitor of GlyT1a (IC50 = 15.8 nM in BE2-C cells expressing the human transporter); selective for GlyT1a over GlyT2 (IC50 = >30 µM in RD cells expressing the human transporter); induces locomotor hyperactivity, impaired gait, compulsive walking, and labored breathing, as well as impairs the ability to stand, in rats at 10 mg/kg; prevents maximal electroshock-induced acute seizures in mice at 30 mg/kg and decreases seizure number in an electroshock-induced rat model of chronic epilepsy at 10 mg/kg