Learn More
Medchemexpress LLC LY2389575 hydrochlor 5mg | 885104-09-6 | 5MG
SDP
Catalog No. 5000240363
Encompass_Preferred
LY2389575 hydrochloride is a selective and noncompetitive mGlu3 negative allosteric modulator (NAM) with an IC50 value of 190 nM LY2389575 hydrochloride induces an increase in Mrc1 levels LY2389575 hydrochloride also independently amplifies Amyloid beta (A ) toxicity and can be used in study of Alzheimer s disease[1 [2 [3
Catalog No. 50-002-40363 Supplier Medchemexpress LLC Supplier No. HY1075095MG
LY2389575 hydrochloride is a selective and noncompetitive mGlu3 negative allosteric modulator (NAM) with an IC50 value of 190 nM LY2389575 hydrochloride induces an increase in Mrc1 levels LY2389575 hydrochloride also independently amplifies Amyloid beta (A ) toxicity and can be used in study of Alzheimer s disease[1 [2 [3