Learn More
Medchemexpress LLC Ly2389575 hydrochloride | 885104-09-6 | 99.9% | 438.58 | C15H16BrCl3N4 | 1 ML
SDP
Catalog No. 5000241329
Encompass_Preferred
LY2389575 hydrochloride is a selective, noncompetitive negative allosteric modulator of the metabotropic glutamate receptor 3 (mGlu3), supplied as a 10 mM solution in DMSO for research use. The compound is reported with high purity and recommended storage conditions for powder and solution forms.
  • Selective, noncompetitive mGlu3 negative allosteric modulator.
  • Provided as a 10 mM solution in DMSO, 1 mL volume.
  • High reported purity suitable for in vitro research applications.
  • Recommended storage conditions specified for powder and solution forms.
  • Useful for studies of glutamatergic signaling and disease-related pathways.
Catalog No. 50-002-41329 Supplier Medchemexpress LLC Supplier No. HY10750910MM 1ML
LY2389575 hydrochloride is a selective, noncompetitive negative allosteric modulator of the metabotropic glutamate receptor 3 (mGlu3), supplied as a 10 mM solution in DMSO for research use. The compound is reported with high purity and recommended storage conditions for powder and solution forms.
  • Selective, noncompetitive mGlu3 negative allosteric modulator.
  • Provided as a 10 mM solution in DMSO, 1 mL volume.
  • High reported purity suitable for in vitro research applications.
  • Recommended storage conditions specified for powder and solution forms.
  • Useful for studies of glutamatergic signaling and disease-related pathways.