Learn More
Medchemexpress LLC LY2389575 hydrochloride | 885104-09-6 | MFCD09745778 | 99.9% | 438.58 g/mol | C15H16BrCl3N4 | 10 MG
SDP
Catalog No. 5000240891
Encompass_Preferred
LY2389575 hydrochloride is the hydrochloride salt of a selective, noncompetitive negative allosteric modulator of the metabotropic glutamate receptor 3 (mGlu3), provided as a high-purity research reagent for preclinical studies, including investigation of amyloid beta-related mechanisms and Alzheimer's disease models.
  • Selective, noncompetitive mGlu3 negative allosteric modulator.
  • High purity (99.9%).
  • Molecular formula C15H16BrCl3N4 and molecular weight 438.58 g/mol.
  • Available in small research quantities, including 10 mg packaging.
  • Stable as powder at -20°C (3 years) or 4°C (2 years); limited stability in solvent.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-40891 Supplier Medchemexpress LLC Supplier No. HY10750910MG
LY2389575 hydrochloride is the hydrochloride salt of a selective, noncompetitive negative allosteric modulator of the metabotropic glutamate receptor 3 (mGlu3), provided as a high-purity research reagent for preclinical studies, including investigation of amyloid beta-related mechanisms and Alzheimer's disease models.
  • Selective, noncompetitive mGlu3 negative allosteric modulator.
  • High purity (99.9%).
  • Molecular formula C15H16BrCl3N4 and molecular weight 438.58 g/mol.
  • Available in small research quantities, including 10 mg packaging.
  • Stable as powder at -20°C (3 years) or 4°C (2 years); limited stability in solvent.
  • For research use only; not for human or clinical use.