Learn More
Medchemexpress LLC HY-114530 10mg Medchemexpress, LY2452473 CAS:1029692-15-6 Purity:>98%
SDP
Catalog No. 501969366
Encompass_Preferred
Medchemexpress, HY-114530 10mg LY2452473 CAS:1029692-15-6 LY2452473 is an orally bioavailable, selective androgen receptor modulator (SARM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9366 Supplier Medchemexpress LLC Supplier No. HY11453010MG
Medchemexpress, HY-114530 10mg LY2452473 CAS:1029692-15-6 LY2452473 is an orally bioavailable, selective androgen receptor modulator (SARM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.