Learn More
Selleck Chemical LLC LY2510924
SDP
Catalog No. 5000000035
Encompass_Preferred
LY2510924 is a potent and selective CXCR4 antagonist that specifically blocks SDF-1 binding to CXCR4 with IC50 value of 0 079 nmol/L and inhibits SDF-1-induced GTP binding with Kb value of 0 38 nmol/L
Catalog No. 50-000-00035 Supplier Selleck Chemical LLC Supplier No. S85055MG
LY2510924 is a potent and selective CXCR4 antagonist that specifically blocks SDF-1 binding to CXCR4 with IC50 value of 0 079 nmol/L and inhibits SDF-1-induced GTP binding with Kb value of 0 38 nmol/L