Learn More
Medchemexpress LLC HY-103704 5mg Medchemexpress, LY2562175 CAS:1103500-20-4 Purity:>98%
SDP
Catalog No. 501968166
Encompass_Preferred
Medchemexpress, HY-103704 5mg LY2562175 CAS:1103500-20-4 LY2562175 is a potent and selective FXR agonist with an EC50 of 193 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8166 Supplier Medchemexpress LLC Supplier No. HY1037045MG
Medchemexpress, HY-103704 5mg LY2562175 CAS:1103500-20-4 LY2562175 is a potent and selective FXR agonist with an EC50 of 193 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.