Learn More
Apexbio Technology LLC LY2584702 1082949-67-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062684
Encompass_Preferred
LY2584702 (CAS 1082949-67-4) is an orally bioavailable ATP-competitive inhibitor selectively targeting p70 S6 kinase (p70S6K) a serine/threonine kinase downstream of the PI3K/Akt/mTOR signaling cascade By inhibiting p70S6K activity LY2584702 effectively suppresses phosphorylation of ribosomal protein S6 (S6) thereby modulating protein translation LY2584702 exhibits potent inhibition of p70S6K (IC50 4 nM) and effectively reduces phosphorylated S6 levels in HCT116 colorectal cancer cells (IC50 between 0 1-0 24 M) Notably LY2584702 demonstrated anticancer activity in preclinical xenograft models supporting its role in targeted cancer research and therapeutic development
Catalog No. 50-000-62684 Supplier Apexbio Technology LLC Supplier No. B58155.1
LY2584702 (CAS 1082949-67-4) is an orally bioavailable ATP-competitive inhibitor selectively targeting p70 S6 kinase (p70S6K) a serine/threonine kinase downstream of the PI3K/Akt/mTOR signaling cascade By inhibiting p70S6K activity LY2584702 effectively suppresses phosphorylation of ribosomal protein S6 (S6) thereby modulating protein translation LY2584702 exhibits potent inhibition of p70S6K (IC50 4 nM) and effectively reduces phosphorylated S6 levels in HCT116 colorectal cancer cells (IC50 between 0 1-0 24 M) Notably LY2584702 demonstrated anticancer activity in preclinical xenograft models supporting its role in targeted cancer research and therapeutic development