Learn More
Selleck Chemical LLC LY2584702 Tosylate
SDP
Catalog No. 5000000394
Encompass_Preferred
LY2584702 Tosylate is a selective ATP-competitive p70S6K inhibitor with IC50 of 4 nM Phase 1
Catalog No. 50-000-00394 Supplier Selleck Chemical LLC Supplier No. S77041G
LY2584702 Tosylate is a selective ATP-competitive p70S6K inhibitor with IC50 of 4 nM Phase 1