Learn More
Selleck Chemical LLC LY2584702 Tosylate 10mg 1082949-68-5
SDP
Supplier:  Selleck Chemical LLC S770410MG
Encompass_Preferred
LY2584702 Tosylate is a selective, ATP-competitive p70S6K inhibitor with IC50 of 4 nM. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0951657