Learn More
Apexbio Technology LLC LY2603618, 10mM (in 1mL DMSO) CAS# 911222-45-2
SDP
Catalog No. 501998393
Encompass_Preferred
LY2603618 is a novel small molecular checkpoint kinase 1 (Chk1) inhibitor that has direct anti-tumour effect [1], as well as desensitizing tumour cell against DNA-damaging chemotherapy treatment [2].Checkpoint kinase 1 (Chk1) is a type of kinase that coordinates damaged-DNA repair [1]. Some tumour cells have been found to possess over-regulated populations of Chk1 that desensitize the cell from DNA-targeting chemotherapy, promoting tumour proliferation [3].LY2603618 inhibits Chk1 by competing with ATP molecules [2]. In A549 and H1299 non-small cancer cell lines, lethal concentration of LY2603618 (10 M) not only resulted in cell proliferation arrest (increase population of cell at G2/M phase from 13% to 38%) but also directly DNA damage, the latter of which was indicated by the increasing occurrence of H2AX phosphorylation[1]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8393 Supplier Apexbio Technology LLC Supplier No. A86385.1
LY2603618 is a novel small molecular checkpoint kinase 1 (Chk1) inhibitor that has direct anti-tumour effect [1], as well as desensitizing tumour cell against DNA-damaging chemotherapy treatment [2].Checkpoint kinase 1 (Chk1) is a type of kinase that coordinates damaged-DNA repair [1]. Some tumour cells have been found to possess over-regulated populations of Chk1 that desensitize the cell from DNA-targeting chemotherapy, promoting tumour proliferation [3].LY2603618 inhibits Chk1 by competing with ATP molecules [2]. In A549 and H1299 non-small cancer cell lines, lethal concentration of LY2603618 (10 M) not only resulted in cell proliferation arrest (increase population of cell at G2/M phase from 13% to 38%) but also directly DNA damage, the latter of which was indicated by the increasing occurrence of H2AX phosphorylation[1]. Other sizes are also available. Please inqury us for quote.