Learn More
Medchemexpress LLC HY-14720 100mg , LY2603618 IC-83 CAS:911222-45-2 Purity:98%
SDP
Catalog No. 501492918
Encompass_Preferred
Medchemexpress, HY-14720 100mg LY2603618 IC-83 CAS:911222-45-2 Formula:C18H22BrN5O3 IC50: 7 nM (Chk1) Purity:98% LY2603618 is a potent and selective inhibitor of Chk1 protein kinase activity in vitro with IC 50 of 7 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2918 Supplier Medchemexpress LLC Supplier No. HY14720100MG
Medchemexpress, HY-14720 100mg LY2603618 IC-83 CAS:911222-45-2 Formula:C18H22BrN5O3 IC50: 7 nM (Chk1) Purity:98% LY2603618 is a potent and selective inhibitor of Chk1 protein kinase activity in vitro with IC 50 of 7 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.