Learn More
Medchemexpress LLC HY-18174A 100mg , LY2606368 (dihydrochloride) Prexasertib dihydrochloride CAS:1234015-54-3 Purity:98%
SDP
Catalog No. 501492763
Encompass_Preferred
Medchemexpress, HY-18174A 100mg LY2606368 (dihydrochloride) Prexasertib dihydrochloride CAS:1234015-54-3 Formula:C18H21Cl2N7O2 Ki: 0.9 nM (CHK1) IC50: <1 nM (CHK1), 8 nM (CHK2) Purity:98% LY2606368 dihydrochloride is a potent and selective ATP competitive inhibitor of the Chk1 protein kinase, with IC 50 s of <1 nM and 8 nM for CHK1 and CHK2, respectively, and a K i of 0.9 nM against purified CHK1. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2763 Supplier Medchemexpress LLC Supplier No. HY18174A100MG
Medchemexpress, HY-18174A 100mg LY2606368 (dihydrochloride) Prexasertib dihydrochloride CAS:1234015-54-3 Formula:C18H21Cl2N7O2 Ki: 0.9 nM (CHK1) IC50: <1 nM (CHK1), 8 nM (CHK2) Purity:98% LY2606368 dihydrochloride is a potent and selective ATP competitive inhibitor of the Chk1 protein kinase, with IC 50 s of <1 nM and 8 nM for CHK1 and CHK2, respectively, and a K i of 0.9 nM against purified CHK1. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.