Learn More
Apexbio Technology LLC LY2608204 1234703-40-2 200mg
SDP
Catalog No. 5000087713
Encompass_Preferred
LY2608204 (CAS 1234703-40-2) is a small molecule activator of glucokinase (GK) enhancing its activity as indicated by an EC50 of 42 nM under low glucose conditions In the INS1-E rat insulinoma cell line LY2608204 stimulates glucose metabolism with an EC50 of 579 nM Preclinical studies have demonstrated that administration of LY2608204 leads to dose-dependent reductions in both fasting and postprandial blood glucose levels These properties make LY2608204 a valuable tool for investigating GK modulation and its effects on glucose homeostasis in metabolic disease research
Catalog No. 50-000-87713 Supplier Apexbio Technology LLC Supplier No. B2160200
LY2608204 (CAS 1234703-40-2) is a small molecule activator of glucokinase (GK) enhancing its activity as indicated by an EC50 of 42 nM under low glucose conditions In the INS1-E rat insulinoma cell line LY2608204 stimulates glucose metabolism with an EC50 of 579 nM Preclinical studies have demonstrated that administration of LY2608204 leads to dose-dependent reductions in both fasting and postprandial blood glucose levels These properties make LY2608204 a valuable tool for investigating GK modulation and its effects on glucose homeostasis in metabolic disease research