Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LY266097 hydrochloride | 172895-39-5 | 99.6% | 407.33 g/mol | C21H24Cl2N2O2 | 1 ML
SDP

Catalog No. 5000246602
Encompass_Preferred

A selective 5-HT2B receptor antagonist supplied as the hydrochloride salt for laboratory research applications. It is used in vitro and in vivo to study 5-HT2B-mediated signaling and neurotransmitter modulation. For research use only.

  • High purity, 99.6%.
  • Molecular weight 407.33 g/mol.
  • Molecular formula C21H24Cl2N2O2.
  • Available as 10 mM 1 mL solution in DMSO or as solid vials (5-100 mg).
  • Recommended storage: sealed at -80°C up to 6 months or at -20°C up to 1 month.

Catalog No. 50-002-46602 Supplier Medchemexpress LLC Supplier No. HY10309410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A selective 5-HT2B receptor antagonist supplied as the hydrochloride salt for laboratory research applications. It is used in vitro and in vivo to study 5-HT2B-mediated signaling and neurotransmitter modulation. For research use only.

  • High purity, 99.6%.
  • Molecular weight 407.33 g/mol.
  • Molecular formula C21H24Cl2N2O2.
  • Available as 10 mM 1 mL solution in DMSO or as solid vials (5-100 mg).
  • Recommended storage: sealed at -80°C up to 6 months or at -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.