Learn More
Medchemexpress LLC Ly266097 hydrochloride | 172895-39-5 | 99.6% | 407.33 g/mol | C21H24Cl2N2O2 | 5 MG
SDP
Catalog No. 5000246374
Encompass_Preferred
LY266097 hydrochloride is a research-grade selective 5-HT2B receptor antagonist supplied as the hydrochloride salt. It is a high-purity solid used in pharmacology and neuropharmacology studies to probe serotonin 5-HT2 receptor signaling and related behavioral and biochemical effects.
  • Selective 5-HT2B antagonist with high affinity.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a stable solid for controlled storage.
  • Well-characterized identifiers: CAS 172895-39-5; molecular weight 407.33 g/mol.
  • Storage conditions compatible with long-term sample integrity.
Catalog No. 50-002-46374 Supplier Medchemexpress LLC Supplier No. HY1030945MG
LY266097 hydrochloride is a research-grade selective 5-HT2B receptor antagonist supplied as the hydrochloride salt. It is a high-purity solid used in pharmacology and neuropharmacology studies to probe serotonin 5-HT2 receptor signaling and related behavioral and biochemical effects.
  • Selective 5-HT2B antagonist with high affinity.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a stable solid for controlled storage.
  • Well-characterized identifiers: CAS 172895-39-5; molecular weight 407.33 g/mol.
  • Storage conditions compatible with long-term sample integrity.