Learn More
Medchemexpress LLC Ly266097 hydrochloride | 172895-39-5 | MFCD00949080 | >98.0% | 407.33 g·mol⁻¹ | C21H23ClN2O2 · HCl | 25 MG
SDP
Catalog No. 5000245994
Encompass_Preferred
LY266097 hydrochloride is the hydrochloride salt of LY-266,097, a potent and selective 5-HT2B receptor antagonist used as a research reagent in neuroscience and pharmacology. It is supplied as a crystalline solid, typically at high purity, and is used for receptor binding and functional studies in vitro and in vivo.
  • Selective 5-HT2B receptor antagonist for neuroscience research.
  • High purity (typically ≥98%) for analytical and biological assays.
  • Available as powder or as a 10 mM solution in DMSO, in milligram-scale packs.
  • Molecular weight ~407.33 g·mol⁻¹; empirical formula C21H23ClN2O2 · HCl.
  • Suited for receptor binding, functional assays, and pharmacology studies.
Catalog No. 50-002-45994 Supplier Medchemexpress LLC Supplier No. HY10309425MG
LY266097 hydrochloride is the hydrochloride salt of LY-266,097, a potent and selective 5-HT2B receptor antagonist used as a research reagent in neuroscience and pharmacology. It is supplied as a crystalline solid, typically at high purity, and is used for receptor binding and functional studies in vitro and in vivo.
  • Selective 5-HT2B receptor antagonist for neuroscience research.
  • High purity (typically ≥98%) for analytical and biological assays.
  • Available as powder or as a 10 mM solution in DMSO, in milligram-scale packs.
  • Molecular weight ~407.33 g·mol⁻¹; empirical formula C21H23ClN2O2 · HCl.
  • Suited for receptor binding, functional assays, and pharmacology studies.