Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ly266097 hydrochloride | 172895-39-5 | MFCD00949080 | 99.6% | 407.33 | C21H24Cl2N2O2 | 10 MG
SDP

Catalog No. 5000246159
Encompass_Preferred

LY266097 hydrochloride is a selective 5-HT2B receptor antagonist supplied as the hydrochloride salt for laboratory research. It is used to study serotonin 5-HT2 receptor pharmacology and is provided at high purity for in vitro and preclinical applications.

  • Selective 5-HT2B receptor antagonist.
  • Reported pKi values: 5-HT2A 7.7; 5-HT2B 9.8; 5-HT2C 7.6.
  • High purity (99.6%) suitable for research use.
  • Supplied as a hydrochloride salt in small milligram quantities for laboratory studies.
  • Molecular formula C21H24Cl2N2O2; molecular weight 407.33.

Catalog No. 50-002-46159 Supplier Medchemexpress LLC Supplier No. HY10309410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY266097 hydrochloride is a selective 5-HT2B receptor antagonist supplied as the hydrochloride salt for laboratory research. It is used to study serotonin 5-HT2 receptor pharmacology and is provided at high purity for in vitro and preclinical applications.

  • Selective 5-HT2B receptor antagonist.
  • Reported pKi values: 5-HT2A 7.7; 5-HT2B 9.8; 5-HT2C 7.6.
  • High purity (99.6%) suitable for research use.
  • Supplied as a hydrochloride salt in small milligram quantities for laboratory studies.
  • Molecular formula C21H24Cl2N2O2; molecular weight 407.33.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.