Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LY266097 hydrochloride | 172895-39-5 | MFCD00949080 | 99.6% | 407.33 | C21H24Cl2N2O2 | 50 MG
SDP

Catalog No. 5000246018
Encompass_Preferred

LY266097 hydrochloride is the hydrochloride salt of LY266097, a selective serotonin 5-HT2B receptor antagonist used in pharmacological research. It is supplied as a solid powder with reported molecular formula C21H24Cl2N2O2, molecular weight 407.33, and high purity suitable for in vitro and in vivo studies. Reported binding affinities include pKi values of 9.8 (5-HT2B), 7.7 (5-HT2A), and 7.6 (5-HT2C).

  • High purity (99.6%).
  • Solid powder, light yellow to light brown.
  • Molecular formula C21H24Cl2N2O2; molecular weight 407.33.
  • Selective 5-HT2B receptor antagonist (reported pKi 9.8 for 5-HT2B).
  • Storage: store sealed at 4°C, away from moisture and light; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use.

Catalog No. 50-002-46018 Supplier Medchemexpress LLC Supplier No. HY10309450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY266097 hydrochloride is the hydrochloride salt of LY266097, a selective serotonin 5-HT2B receptor antagonist used in pharmacological research. It is supplied as a solid powder with reported molecular formula C21H24Cl2N2O2, molecular weight 407.33, and high purity suitable for in vitro and in vivo studies. Reported binding affinities include pKi values of 9.8 (5-HT2B), 7.7 (5-HT2A), and 7.6 (5-HT2C).

  • High purity (99.6%).
  • Solid powder, light yellow to light brown.
  • Molecular formula C21H24Cl2N2O2; molecular weight 407.33.
  • Selective 5-HT2B receptor antagonist (reported pKi 9.8 for 5-HT2B).
  • Storage: store sealed at 4°C, away from moisture and light; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.