Learn More
Cayman Chemical LY266097 (hydrochloride), Purity: ≥98%, CAS: 172895-39-5, 10 mg
SDP
Catalog No. 502953283
Encompass_Preferred
A 5-HT2B receptor antagonist (Ki = 0.5 nM); selective for 5-HT2B over 5-HT2A and 5-HT2C receptors (Kis = 19.5 and 24.55 nM, respectively); reduces L5/L6 spinal nerve ligation-induced hind paw tactile allodynia in a rat model of neuropathic pain at an intrathecal dose of 100 nmol; reduces hyperlocomotion induced by cocaine in rats at 0.63 mg/kg
Catalog No. 50-295-3283 Supplier Cayman Chemical Supplier No. 1193510 MG
A 5-HT2B receptor antagonist (Ki = 0.5 nM); selective for 5-HT2B over 5-HT2A and 5-HT2C receptors (Kis = 19.5 and 24.55 nM, respectively); reduces L5/L6 spinal nerve ligation-induced hind paw tactile allodynia in a rat model of neuropathic pain at an intrathecal dose of 100 nmol; reduces hyperlocomotion induced by cocaine in rats at 0.63 mg/kg