Learn More
Medchemexpress LLC HY-13034 100mg , LY2784544 gandotinib CAS:1229236-86-5 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13034100MG
Encompass_Preferred
Medchemexpress, HY-13034 100mg LY2784544 gandotinib CAS:1229236-86-5 Formula:C23H25ClFN7O IC50: 2.52±0.49 nM (JAK2), 19.8±3 nM (JAK1), 48±14.9 nM (JAK3), 4 nM (FLT3), 32 nM (FGFR2) Purity:98% LY2784544 is a potent JAK2 inhibitor with IC 50 of 3 nM. LY2784544 also inhibits FLT3, FLT4, FGFR2, TYK2, and TRKB with IC 50 of 4, 25, 32, 44, and 95 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2958