Learn More
Selleck Chemical LLC Gandotinib (LY2784544) 50mg 1229236-86-5
SDP
Supplier:  Selleck Chemical LLC S217950MG
Encompass_Preferred
Gandotinib (LY2784544) is a potent JAK2 inhibitor with IC50 of 3 nM, effective in JAK2V617F, 8- and 20-fold selective versus JAK1 and JAK3. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-849-3