Learn More
Medchemexpress LLC HY-119243 10mg Medchemexpress, LY2794193 CAS:2173037-97-1 Purity:>98%
SDP
Catalog No. 501969521
Encompass_Preferred
Medchemexpress, HY-119243 10mg LY2794193 CAS:2173037-97-1 LY2794193 is a highly potent and selective mGlu3 receptor agonist (hmGlu3 Ki=0.927 nM,EC50=0.47 nM; hmGlu2 Ki=412 nM,EC50=47.5 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9521 Supplier Medchemexpress LLC Supplier No. HY11924310MG
Medchemexpress, HY-119243 10mg LY2794193 CAS:2173037-97-1 LY2794193 is a highly potent and selective mGlu3 receptor agonist (hmGlu3 Ki=0.927 nM,EC50=0.47 nM; hmGlu2 Ki=412 nM,EC50=47.5 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.