Learn More
Medchemexpress LLC HY-119243 5mg Medchemexpress, LY2794193 CAS:2173037-97-1 Purity:>98%
SDP
Catalog No. 501932534
Encompass_Preferred
Medchemexpress, HY-119243 5mg LY2794193 CAS:2173037-97-1 LY2794193 is a highly potent and selective mGlu3 receptor agonist ( hmGlu3 K i =0.927 nM, EC 50 =0.47 nM; hmGlu2 K i =412 nM, EC 50 =47.5 nM) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2534 Supplier Medchemexpress LLC Supplier No. HY1192435MG
Medchemexpress, HY-119243 5mg LY2794193 CAS:2173037-97-1 LY2794193 is a highly potent and selective mGlu3 receptor agonist ( hmGlu3 K i =0.927 nM, EC 50 =0.47 nM; hmGlu2 K i =412 nM, EC 50 =47.5 nM) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.