Learn More
Medchemexpress LLC HY-18760 1mg Medchemexpress, LY2812223 CAS:1311385-20-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY187601MG
Encompass_Preferred
Medchemexpress, HY-18760 1mg LY2812223 CAS:1311385-20-2 LY2812223 is a highly potent, functionally selective mGlu2 receptor agonist with mGlu2 binding affinity for mGlu2 and mGlu3 (Ki=144 nM and 156 nM, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0013