Learn More
Medchemexpress LLC Abemaciclib methanesulfonate, 1231930-82-7, MFCD25562906, 500mg
SDP
Supplier:  Medchemexpress LLC HY16297500MG
Encompass_Preferred
Formula C₂₈H₃₆F₂N₈O₃S, Molecular Weight 602.70, SMILES: CC(N1C2=CC(C3=NC(NC4=NC=C(CN5CCN(CC)CC5)C=C4)=NC=C3F)=CC(F)=C2N=C1C)C.CS(=O)(O)=O; Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.
Catalog No. 50-163-8512