Learn More
Apexbio Technology LLC LY2835219 1231930-82-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062849
Encompass_Preferred
LY2835219 (CAS 1231930-82-7) is a selective orally bioavailable inhibitor of cyclin-dependent kinases 4 and 6 (CDK4/6) It targets CDK4 and CDK6 with reported IC50 values of 2 nM and 10 nM respectively thereby reducing phosphorylation of the retinoblastoma protein (Rb) This mechanism leads to cell-cycle arrest during the G1 phase and subsequent inhibition of tumor cell proliferation LY2835219 demonstrates the capacity to cross the blood-brain barrier indicating its potential utility in research involving primary or metastatic intracranial tumors
Catalog No. 50-000-62849 Supplier Apexbio Technology LLC Supplier No. A17945.1
LY2835219 (CAS 1231930-82-7) is a selective orally bioavailable inhibitor of cyclin-dependent kinases 4 and 6 (CDK4/6) It targets CDK4 and CDK6 with reported IC50 values of 2 nM and 10 nM respectively thereby reducing phosphorylation of the retinoblastoma protein (Rb) This mechanism leads to cell-cycle arrest during the G1 phase and subsequent inhibition of tumor cell proliferation LY2835219 demonstrates the capacity to cross the blood-brain barrier indicating its potential utility in research involving primary or metastatic intracranial tumors