Learn More
AdipoGen LY2835219
SDP
Catalog No. 502560597
Encompass_Preferred
Chemical. CAS 1231930-82-7. Formula C27H32F2N8 . CH4O3S. MW 602.7. LY2835219 is an orally-bioavailable dual inhibitor of cyclin-dependent kinase 4 CDK4 and CDK6 IC50s = 2 and 10 nM, respectively. Through this mechanism, it blocks phosphorylation of retinoblastoma protein, resulting in arrest of cell cycling in the G1 phase. LY2835219 has antitumor action against xenografts when used alone or in combination with other chemotherapeutic compounds.
Catalog No. 50-256-0597 Supplier AdipoGen Supplier No. CDXL0303M250
Chemical. CAS 1231930-82-7. Formula C27H32F2N8 . CH4O3S. MW 602.7. LY2835219 is an orally-bioavailable dual inhibitor of cyclin-dependent kinase 4 CDK4 and CDK6 IC50s = 2 and 10 nM, respectively. Through this mechanism, it blocks phosphorylation of retinoblastoma protein, resulting in arrest of cell cycling in the G1 phase. LY2835219 has antitumor action against xenografts when used alone or in combination with other chemotherapeutic compounds.