Learn More
Selleck Chemical LLC Abemaciclib mesylate (LY2835219) 5mg 1231930-82-7
SDP
Catalog No. NC0577560
Encompass_Preferred
Abemaciclib (LY2835219) is a potent and selective inhibitor of CDK4 and CDK6 with IC50 of 2 nM and 10 nM in cell-free assays, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0577560 Supplier Selleck Chemical LLC Supplier No. S71585MG
Abemaciclib (LY2835219) is a potent and selective inhibitor of CDK4 and CDK6 with IC50 of 2 nM and 10 nM in cell-free assays, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.