Learn More
Medchemexpress LLC HY-16297A 100mg Medchemexpress, LY2835219 (free base) Abemaciclib;CDK4/6 dual inhibitor CAS:1231929-97-7 Purity:98%
SDP
Catalog No. 501638513
Encompass_Preferred
Medchemexpress,HY-16297A 100mg LY2835219 (free base) Abemaciclib;CDK4/6 dual inhibitor CAS:1231929-97-7 Formula:C27H32F2N8 LY2835219 free base is a selective CDK4/6 inhibitor with IC50 values of 2 nM and 10 nM for CDK4 and CDK6, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8513 Supplier Medchemexpress LLC Supplier No. HY16297A100MG
Medchemexpress,HY-16297A 100mg LY2835219 (free base) Abemaciclib;CDK4/6 dual inhibitor CAS:1231929-97-7 Formula:C27H32F2N8 LY2835219 free base is a selective CDK4/6 inhibitor with IC50 values of 2 nM and 10 nM for CDK4 and CDK6, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.