Learn More
Cayman Chemical LY2835219 (methanesulfonate), Purity: ≥95%, CAS: 1231930-82-7, 10 mg
SDP
Catalog No. 502968186
Encompass_Preferred
An orally-bioavailable dual inhibitor of CDK4 and CDK6 (IC50s = 2 and 10 nM, respectively); has antitumor action against xenografts when used alone or in combination with other chemotherapeutic compounds
Catalog No. 50-296-8186 Supplier Cayman Chemical Supplier No. 1774010 MG
An orally-bioavailable dual inhibitor of CDK4 and CDK6 (IC50s = 2 and 10 nM, respectively); has antitumor action against xenografts when used alone or in combination with other chemotherapeutic compounds