Learn More
Selleck Chemical LLC LY2857785
SDP
Catalog No. 5000002868
Encompass_Preferred
LY2857785 is a type I reversible and competitive ATP kinase inhibitor against CDK9(IC50 0 011 M) and also inhibits other transcription kinases CDK8(IC50 0 016 M) and CDK7 (IC50 0 246 M)
Catalog No. 50-000-02868 Supplier Selleck Chemical LLC Supplier No. S75112MG
LY2857785 is a type I reversible and competitive ATP kinase inhibitor against CDK9(IC50 0 011 M) and also inhibits other transcription kinases CDK8(IC50 0 016 M) and CDK7 (IC50 0 246 M)