Learn More
Medchemexpress LLC HY-12293 5mg Medchemexpress, LY2857785 CAS:1619903-54-6 Purity:>98%
SDP
Catalog No. 502029943
Encompass_Preferred
Medchemexpress, HY-12293 5mg LY2857785 CAS:1619903-54-6 LY2857785 is a type I reversible and competitive ATP kinase inhibitor against CDK9 (IC50 11 nM) and other transcription kinases CDK8 (IC50 16 nM), and CDK7 (IC50 246 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9943 Supplier Medchemexpress LLC Supplier No. HY122935MG
Medchemexpress, HY-12293 5mg LY2857785 CAS:1619903-54-6 LY2857785 is a type I reversible and competitive ATP kinase inhibitor against CDK9 (IC50 11 nM) and other transcription kinases CDK8 (IC50 16 nM), and CDK7 (IC50 246 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.